Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylformamide ≥99.8% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DimethylformamideDMFDMFAFormic acid dimethylamideFormyldimethylamineN-Formyldimethylamine

039117-M1 39117-M6 039117-K7 39117-K2 39117-AP 039117-K2 39117-K7 039117-AP 39117-M1 39117-M6
AA39117-APEA 76.4 USD InStock
CAAA39117-M1 AA39117-AP AA39117-K7 CAAA39117-AP CAAA39117-K7 AA39117-M6 AA39117-K2 CAAA39117-M6 AA39117-M1 CAAA39117-K2
N,N-Dimethylformamide ≥99.8% ACS
N,N-Dimethylformamide
A solvent for chromogenic substrates


Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C
Melting Pt: –61 °C
Density: 0.944
Flash Pt: 57 °C (135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg
Order Now
Specification Test Results
Appearance Pass test
Color (APHA) Pass test.
Evaporation residue 0.005% max.
Titratable base 0.003meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty