Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium niobium methoxide 5% (w/v) in methanol , 99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

39154-AC 039154-AC 39154-AE 039154-AE
AA39154-ACEA 84.29 USD InStock
AA39154-AC CAAA39154-AE AA39154-AE CAAA39154-AC
Lithium niobium methoxide 5% (w/v) in methanol , 99% (metals basis)
Lithium niobium methoxide


Formula: C₆H₁₈LiNbO₆
MW: 285.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210627
CAS Number: 21864-11-9
UN: 1230
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty