Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium-2-ethylhexanoate solution ≥75% (w/w)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Ethylhexanoic Acid Potassium Salt

039163-04 039163-18 39163-18 39163-04
AA39163-04EA 35.69 USD InStock
CAAA39163-04 CAAA39163-18 AA39163-04 AA39163-18
Potassium-2-ethylhexanoate solution ≥75% (w/w)
Potassium-2-ethylhexanoate
Contains a mixture of organic solvents


Formula: C₈H₁₆O₂
MW: 182.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045896
CAS Number: 3164-85-0
EINECS: 221-625-7
UN: 1993
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty