Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) 1,1,1-trifluoro-2,4-pentanedionate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tris(trifluoro-2,4-pentanedionato)iron(III)Ferric(III) trifluoroacetylacetonateIron(III) trifluoroacetylacetonateTrifluoroacetylacetono Iron(III) salt

039265-06 39265-06 039265-14 39265-14
AA39265-06EA 164.99 USD InStock
AA39265-06 CAAA39265-14 AA39265-14 CAAA39265-06
Iron(III) 1,1,1-trifluoro-2,4-pentanedionate
Iron(III) 1,1,1-trifluoro-2,4-pentanedionate


Formula: C₁₅H₁₂F₉FeO₆
MW: 515.086 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00017760
CAS Number: 14526-22-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty