Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%

Supplier: Thermo Scientific Chemicals
Danger

39303-03 039303-MD 039303-03 39303-MD
AA39303-03EA 151.99 USD InStock
CAAA39303-03 AA39303-03 CAAA39303-MD AA39303-MD
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer


Formula: C₂₄H₃₀Mo₂O₄
Melting Pt: 219 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00058846
CAS Number: 12132-04-6
UN: 3466
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty