Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(isopropylcyclopentadienyl)lanthanum(III) ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tris(i-propylcyclopentadienyl)lanthanum

039323-03
AA39323-03EA 123.99 USD InStock
AA39323-03
Tris(isopropylcyclopentadienyl)lanthanum(III) ≥99.9% (REO, rare earth oxide basis)
Tris(isopropylcyclopentadienyl)lanthanum(III)


Formula: C₂₄H₃₃La
MW: 460.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145482
CAS Number: 68959-87-5
UN: 3181
ADR: ,
Order Now
Specification Test Results
Assay Minimum 99.9

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty