Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium stearate

Supplier: Thermo Scientific Chemicals
Synonyms: Stearic acid lithium salt

39335-A1 039335-30 39335-30 039335-A1
AA39335-30EA 62.79 USD InStock
AA39335-30 CAAA39335-30 AA39335-A1 CAAA39335-A1
Lithium stearate
Lithium stearate


Formula: C₁₈H₃₅LiO₂
MW: 290.42 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00042032
CAS Number: 4485-12-5
EINECS: 224-772-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty