Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) metaborate monohydrate

Supplier: Thermo Scientific Chemicals
Danger

039339-22 39339-14 39339-22 39339-14
AA39339-14EA 96.17 USD InStock
AA39339-14 CAAA39339-22 CAAA39339-14 AA39339-22
Lead(II) metaborate monohydrate
Lead(II) metaborate monohydrate
Insoluble in water and alkali. Soluble in acid


Formula: B₂O₄Pb.H₂O
MW: 310.81 g/mol (292.81 anhy)
Melting Pt: 160 °C (-H₂O)
Density: 5,598
Storage Temperature: Ambient
MDL Number: MFCD00049504
CAS Number: 10214-39-8
EINECS: 238-762-3
UN: 2291
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty