Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium orthoarsenate 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Lithium arsenate

39350-06 39350-14 039350-14 039350-06
AA39350-14EA 164.99 USD InStock
CAAA39350-06 AA39350-14 AA39350-06 CAAA39350-14
Lithium orthoarsenate 99%
Lithium orthoarsenate


Formula: AsLi₃O₄
MW: 159.74 g/mol
Density: 3,07
Storage Temperature: Ambient
MDL Number: MFCD00053998
CAS Number: 13478-14-3
EINECS: 236-773-8
UN: 1557
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty