Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) oxalate dihydrate

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nickel oxalate dihydrate

039454-30 039454-18 39454-30 39454-18
AA39454-18EA 135.99 USD InStock
AA39454-18 CAAA39454-18 CAAA39454-30 AA39454-30
Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate
Soluble in acids. Insoluble in water


Formula: C₂H₄NiO₆
MW: 182.741 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167155
CAS Number: 6018-94-6
EINECS: 208-933-7
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty