Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) cyanide tetrahydrate

Supplier: Thermo Scientific Chemicals
Danger

39480-18 039480-18 39480-09 039480-09
AA39480-18EA 835.99 USD InStock
CAAA39480-18 AA39480-18 CAAA39480-09 AA39480-09
Nickel(II) cyanide tetrahydrate
Nickel(II) cyanide tetrahydrate
Applications: Metallurgy, nickel-plating
Insoluble in water. Soluble in ammonium carbonate, ammonia, alkali cyanides


Formula: Ni(CN)₂.4H₂O
MW: 182.73 g/mol (110.73 anhy)
Melting Pt: 200 °C (-4H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00049490
CAS Number: 13477-95-7
EINECS: 209-160-8
UN: 1653
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty