Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium hydroxide on carbon ≥99% (Pearlman's catalyst)

Supplier: Thermo Scientific Chemicals
Synonyms: Pearlman's Catalyst (contains PdPdO)

39496-01 39496-06 039496-03 39496-03 039496-01 039496-06
AA39496-06EA 1160.99 USD InStock
AA39496-06 AA39496-03 CAAA39496-06 CAAA39496-01 CAAA39496-03 AA39496-01
Palladium hydroxide on carbon ≥99% (Pearlman's catalyst)
Palladium hydroxide


Formula: H₂O₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS Number: 12135-22-7
EINECS: 235-219-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty