Would you like to log in again?
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), wire, annealed, Diameter 0.2 mm (0.008 in)
Supplier: Thermo Scientific Chemicals
039526-BW 39526-BS 039526-BS
AA39526-BWEA 496.49 USD InStock
AA39526-BW CAAA39526-BW AA39526-BS CAAA39526-BS
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), wire, annealed, Diameter 0.2 mm (0.008 in)
Platinum Iridium alloy (80:20 w%)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty