Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium(IV) oxide-2-ethylhexanoate (∼6% Zr) in mineral spirits

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Zirconyl 2-ethylhexanoate

039543-36 39543-36
AA39543-36EA 205.99 USD InStock
AA39543-36 CAAA39543-36
Zirconium(IV) oxide-2-ethylhexanoate (∼6% Zr) in mineral spirits
Zirconium(IV) oxide-2-ethylhexanoate


Formula: (C₈H₁₅O₂)xZn
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00072684
CAS Number: 22464-99-9
UN: 1268
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty