Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc naphthenate 67% (10% Zn) in mineral spirits

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Naphthenic Acid Zinc Salt

39554-36 39554-A3 039554-36 039554-A3
AA39554-A3EA 262.99 USD InStock
CAAA39554-A3 CAAA39554-36 AA39554-A3 AA39554-36
Zinc naphthenate 67% (10% Zn) in mineral spirits
Zinc naphthenate


Formula: 2(C₁₁H₇O₂)·Zn
MW: 319.71 g/mol
Density: 1,01
Flash Pt: >37.8° (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00147699
CAS Number: 12001-85-3
UN: 1993
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty