Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanadium(IV) oxide meso-tetraphenylporphine

Supplier: Thermo Scientific Chemicals
Synonyms: Vanadyl meso-tetraphenylporphine

39583-ME 39583-MC 039583-ME
AA39583-MEEA 197.99 USD InStock
CAAA39583-MC AA39583-ME CAAA39583-ME
Vanadium(IV) oxide meso-tetraphenylporphine
Vanadium(IV) oxide meso-tetraphenylporphine


Formula: C₄₄H₂₈N₄OV
MW: 679.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012154
CAS Number: 14705-63-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty