Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diindium trisulfate, anhydrous ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Synonyms: Diindium tris(sulphate) Indium (III) sulphate

40102-22 040102-22 040102-14 40102-14
AA40102-22EA 597.99 USD InStock
CAAA40102-14 AA40102-22 CAAA40102-22 AA40102-14
Diindium trisulfate, anhydrous ≥99.99% (metals basis)
Diindium trisulphate


Formula: O₁₂In₂S₃
MW: 517.83 g/mol
Melting Pt: 585 °C
Density: 3.438
Storage Temperature: Ambient
MDL Number: MFCD00011059
CAS Number: 13464-82-9
EINECS: 236-689-1
Merck Index: 13,04978
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty