Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium carbide ≥99.5% (metals basis), Powder

Supplier: Thermo Scientific Chemicals
Warning

040178-18 40178-18 040178-30 40178-A1 040178-A1 40178-30
AA40178-18EA 75.69 USD InStock
CAAA40178-18 AA40178-18 AA40178-30 CAAA40178-A1 AA40178-A1 CAAA40178-30
Titanium carbide ≥99.5% (metals basis), Powder
Titanium carbide


Formula: CTi
Boiling Pt: 4820 °C
Melting Pt: ∼ 3140 °C
Density: 4,93
Storage Temperature: Ambient
MDL Number: MFCD00011268
CAS Number: 12070-08-5
EINECS: 235-120-4
UN: 3178
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty