Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iodide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

040255-22 40255-06 40255-22 040255-06 040255-14 40255-14
AA40255-22EA 929.99 USD InStock
AA40255-22 CAAA40255-22 CAAA40255-06 AA40255-06 CAAA40255-14 AA40255-14
Ammonium iodide ≥99.995% (metals basis)
Ammonium iodide


Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551 °C (subl.)
Density: 2.514
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 13,00531
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty