Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium ammonium phosphate tetrahydrate

40320-A4 040320-22 40320-22 040320-36 040320-A4 40320-36
AA40320-22EA 29.92 USD InStock
AA40320-22 CAAA40320-A4 CAAA40320-36 AA40320-36 AA40320-A4 CAAA40320-22
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Sodium ammonium hydrogen phosphate tetrahydrate
Soluble in water. Insoluble in alcohol


Formula: NaNH₄HPO₄·4 H₂O
MW: 209.07 g/mol
Melting Pt: 79 °C (dec.)
Density: 1.544
Storage Temperature: Ambient
MDL Number: MFCD00066970
CAS Number: 7783-13-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty