Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bismuth chloride oxide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Bismuth oxychloride

040396-30 40396-30 040396-A1 040396-18 40396-A1 40396-18
AA40396-18EA 57 USD InStock
AA40396-18 CAAA40396-30 CAAA40396-A1 AA40396-30 AA40396-A1 CAAA40396-18
Bismuth chloride oxide 98%
Bismuth chloride oxide
Used as pigment, in manufacture of artificial pearls, dry-cell cathodes


Formula: BiClO
MW: 260.43 g/mol
Melting Pt: Dec.
Density: 7,72
Storage Temperature: Ambient
MDL Number: MFCD00010895
CAS Number: 7787-59-9
EINECS: 232-122-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty