Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific

Supplier: Thermo Scientific
40447-K2 42417-K7 40439-K2 042416-AP 42418-K2 038712-K7 40446-AP 38713-AP 42421-AP 42418-K7 042421-AP 38711-K7 042424-K2 042417-K2 042420-AP 038714-K7 38709-K2 042425-K7 42420-AP 046307-K2 042425-K2 038712-AP 038712-K2 040447-K2 42423-K7 038710-K7 38709-K7 42418-AP 42423-K2 040446-K2 042420-K2 38714-K7 042419-K2 038713-AP 042422-K2 42415-AP 042422-AP 042415-AP 042425-AP 040439-K2 038709-K7 42425-K7 038709-AP 42422-K7 38711-K2 042418-AP 42420-K7 038711-K2 042421-K2 38713-K2 046307-AP 38710-K2 42424-K7 38712-K7 042417-K7 42415-K7 42416-K7 042420-K7 040439-K7 038710-AP 40447-K7 40439-K7 042423-K2 38713-M4 040439-AP 38711-AP 42417-K2 042416-K2 038709-K2 42422-K2 42425-K2 42419-K2 042423-AP 042417-AP 40446-K7 38714-K2 42421-K2 42420-K2 042415-K2 42425-AP 42421-K7 38710-K7 42415-K2 042421-K7 42424-AP 42423-AP 42416-AP 40446-K2 38713-K7 040446-AP 042424-K7 42419-AP 42416-K2 038713-K2 038710-K2 038713-M4 42423-K7 042419-K7 046307-K7 38712-AP 040446-K7 038714-AP 042424-AP 38714-AP 040447-K7 42419-K7 40447-AP 042422-K7 038711-K7 042415-K7 042418-K7 38712-K2 038711-AP 42424-K2 38709-AP 40439-AP 42422-AP 42417-AP 38710-AP 038714-K2 042419-AP 040447-AP 038713-K7 042418-K2 042416-K7
AA40447-APEA 50.25 USD InStock
CAAA42421-K2 AA40447-AP AA42416-AP AA42422-AP AA38711-AP CAAA42415-K2 CAAA42415-K7 CAAA40446-AP CAAA42418-AP AA42422-K2 CAAA42415-AP CAAA40446-K7 CAAA40446-K2 CAAA42421-AP AA40447-K2 AA42416-K2 CAAA38710-AP AA42416-K7 AA42419-AP AA40447-K7 CAAA42424-K7 AA38713-K2 CAAA42424-K2 AA38713-K7 CAAA42418-K2 CAAA42418-K7 AA42425-AP AA42422-K7 AA42425-K2 CAAA38709-K7 AA42419-K2 CAAA38712-K7 AA42419-K7 CAAA42424-AP CAAA38709-K2 CAAA42421-K7 AA38710-K2 CAAA42417-K2 CAAA42423-K2 CAAA42417-K7 AA38710-K7 AA42421-K7 AA38713-AP AA42424-AP AA40439-AP CAAA38712-K2 CAAA40447-AP AA42424-K2 AA42418-K2 CAAA38712-AP CAAA38709-AP CAAA42423-AP CAAA42420-K7 AA42418-K7 AA40439-K2 AA40439-K7 AA38710-AP AA42424-K7 CAAA38714-K7 CAAA42423-K7 CAAA42425-K7 AA38712-K2 AA46307-K2 AA38712-K7 CAAA42425-K2 CAAA42419-K2 CAAA42419-K7 AA42423-K7 CAAA38714-K2 CAAA38711-K2 CAAA38714-AP CAAA38711-K7 AA46307-AP CAAA42425-AP CAAA42422-K7 CAAA40439-AP AA38709-K2 AA42415-AP CAAA42420-K2 AA38709-K7 CAAA40447-K2 AA42421-AP AA38712-AP CAAA40447-K7 CAAA42417-AP AA42421-K2 AA38709-AP CAAAA38713-M4 AA46307-K7 CAAA38711-AP CAAA42420-AP AA42415-K2 AA42418-AP AA42415-K7 AA38714-K7 AA42420-AP CAAA42416-AP AA42425-K7 CAAA38713-K7 AA38711-K2 CAAA42422-K2 AA42417-AP AA40446-AP AA38714-AP CAAA42416-K2 AA38711-K7 AA42420-K7 AA42423-AP AA42420-K2 CAAA38713-K2 CAAA42416-K7 CAAA42419-AP AA42423-K2 CAAA38710-K2 CAAA38710-K7 AAA38713-M4 CAAA38713-AP CAAA40439-K2 AA42417-K2 AA40446-K2 CAAA40439-K7 CAAA42422-AP AA38714-K2 AA42417-K7 AA40446-K7
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific
Buffers pH Calibration Buffers
NIST Traceable.

Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD03458582
Calibration of pH meters

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty