Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium(III)-2-ethylhexanoate 49% (12% Ce) in 2-ethylhexanoic acid

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Cerium(III) 2-ethylhexanoate

40451-A3 40451-36 040451-22 040451-A3 040451-36 40451-22
AA40451-A3EA 291.99 USD InStock
AA40451-A3 CAAA40451-36 CAAA40451-A3 AA40451-22 AA40451-36 CAAA40451-22
Cerium(III)-2-ethylhexanoate 49% (12% Ce) in 2-ethylhexanoic acid
Cerium (III) 2-ethylhexanoate


Formula: C₂₄H₄₅CeO₆
MW: 569.727 g/mol
Density: 1,08
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014004
CAS Number: 56797-01-4
EINECS: 260-386-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty