Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium(IV) oxide ≥99.4% (metals basis excluding Hf) stabilized, powder -100+325 Mesh

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Zirconium dioxide

Stabilization: Stabilised with calcia (CaO) 4%

40453-30 40453-A1 040453-18 40453-18 040453-A1
AA40453-18EA 44.19 USD InStock
CAAA40453-18 AA40453-18 AA40453-30 CAAA40453-A1 AA40453-A1
Zirconium(IV) oxide ≥99.4% (metals basis excluding Hf) stabilized, powder -100+325 Mesh
Zirconium (IV) oxide
The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.

Plasma Spray Grade. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, Alcohols, Halogens.


Formula: O₂Zr
MW: 123.22 g/mol
Boiling Pt: ∼ 5000 °C
Melting Pt: ∼ 2700 °C
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty