Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) oxide ≥99.9% (metals basis), REacton®, high surface area

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Titanium dioxide Pigment White 6

40458-04 40458-09 040458-09 040458-04
AA40458-09EA 383.99 USD InStock
AA40458-09 CAAA40458-09 AA40458-04 CAAA40458-04
Titanium(IV) oxide ≥99.9% (metals basis), REacton®, high surface area
Titanium (IV) oxide
Photocatalyst, catalyst, catalyst support


Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2500…3000 °C
Melting Pt: 1830…1850 °C
Density: 4,26
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty