Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dirhenium heptaoxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Rhenium (VII) oxide

40468-04 40468-09 040468-09 040468-18 40468-18 040468-04
AA40468-09EA 555.99 USD InStock
AA40468-09 CAAA40468-18 CAAA40468-09 AA40468-18 CAAA40468-04 AA40468-04
Dirhenium heptaoxide ≥99.9% (metals basis)
di-Rhenium heptaoxide
As a catalyst for oxidation/reduction reactions


Formula: O₇Re₂
MW: 484.41 g/mol
Boiling Pt: 360 °C
Melting Pt: 297 °C
Density: 6,103
Storage Temperature: Ambient
MDL Number: MFCD00011199
CAS Number: 1314-68-7
EINECS: 215-241-9
UN: 3085
ADR: 5.1,III
Merck Index: 12,08344
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty