Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Erbium triacetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Synonyms: Erbium(III) acetate tetrahydrate

040481-14 40481-22 40481-06 40481-14 040481-22 040481-06
AA40481-06EA 49.79 USD InStock
AA40481-06 CAAA40481-06 CAAA40481-14 AA40481-14 AA40481-22 CAAA40481-22
Erbium triacetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium triacetate tetrahydrate


Formula: C₆H₉ErO₆·4H₂O
MW: 416.45 g/mol
Density: 2,114
Storage Temperature: Ambient
MDL Number: MFCD00013034
CAS Number: 15280-57-6
EINECS: 247-067-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty