Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ICP-MS Memory Check Standard solution B for 6020 CLP-M, Specpure®, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
040488-AE 040488-AP
AA40488-AEEA 178.99 USD InStock
AA40488-AE AA40488-AP
ICP-MS Memory Check Standard solution B for 6020 CLP-M, Specpure®, Thermo Scientific
Standards Cationic Standards Multi-Element Standards
Molecular Formula: Matrix: 2% HNO3/tr. F-/tr. tartaric acid
Physical Form: Liquid
Density: 1.0
UN No.: 3264
MDL No.: MFCD00803323

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty