Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel (II) sulfamate 50% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nickel bis(sulphamidate)Nickel (II) sulfamidateNickel bis(sulfamidate)Nickel sulphamateNickel (II) sulphamidateNickel sulfamate

40501-K2 040501-K2 040501.8M 40501-8M
AA40501-K2EA 61.49 USD InStock
AA40501-K2 CAAA40501-K2 AAA40501-8M CAAAA40501-8M
Nickel (II) sulfamate 50% (w/w) in aqueous solution, Reagent Grade
Nickel (II) sulphamate
Nickel plating


Formula: H₄N₂NiO₆S₂
MW: 250.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00137261
CAS Number: 13770-89-3
EINECS: 237-396-1
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty