Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylenediaminetetraacetic acid manganese disodium salt hydrate

Supplier: Thermo Scientific Chemicals
Synonyms: Manganese Disodium Ethylenediaminetetraacetate

40519-06 040519-14 040519-06 40519-14
AA40519-14EA 105.99 USD InStock
AA40519-14 CAAA40519-14 CAAA40519-06 AA40519-06
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
Applications: Metal chelating compound

Applications: Metal chelating compound


Formula: C₁₀H₁₆N₂O₈
MW: 389.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00661156
CAS Number: 15375-84-5
EINECS: 239-407-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty