Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Ethylmaleimide (NEM) 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Ethyl-1H-pyrrole-2,5-dione

040526-06 040526-03 40526-03 40526-14 040526-14 40526-06
AA40526-03EA 46.4 USD InStock
CAAA40526-03 CAAA40526-14 AA40526-03 AA40526-14 CAAA40526-06 AA40526-06
N-Ethylmaleimide (NEM) 99+%
N-Ethylmaleimide


Formula: C₆H₇NO₂
MW: 125.13 g/mol
Boiling Pt: 209…211 °C
Melting Pt: 43…46 °C
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005509
CAS Number: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1,II
Merck Index: 13,03855
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty