Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scandium (III) trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Scandium(III) trifluoromethanesulphonateScandium triflateScandium(III) trifluoromethanesulfonate

40566-03 40566-06 40566-MD 040566-03 040566.MD 040566-06
AA40566-03EA 66.89 USD InStock
CAAA40566-03 CAAAA40566-MD AA40566-03 CAAA40566-06 AA40566-06 AAA40566-MD
Scandium (III) trifluoromethanesulfonate 98%
Scandium (III) trifluoromethanesulphonate


Formula: C₃F₉O₉S₃Sc
MW: 492.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00798539
CAS Number: 144026-79-9
Order Now
Specification Test Results
Form Powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty