Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium fluoride ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

040755-04 040755-09 40755-18 40755-04 040755-18 40755-09
AA40755-18EA 825.99 USD InStock
CAAA40755-09 AA40755-18 AA40755-09 AA40755-04 CAAA40755-18 CAAA40755-04
Magnesium fluoride ≥99.99% (metals basis), Puratronic®
Magnesium fluoride


Formula: MgF₂
MW: 62.3 g/mol
Boiling Pt: 2260 °C (1013 hPa)
Melting Pt: 1261 °C
Density: 3.148
Storage Temperature: Ambient
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Merck Index: 12,05701
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty