Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 20 mesh woven from 0.382mm (0.015in) Type 304

Supplier: Thermo Scientific Chemicals

040942-HB 40942-HB 40942-FL 40942-GT 040942-GT 040942-FL
AA40942-GTEA 64.09 USD InStock
CAAA40942-FL AA40942-GT AA40942-HB CAAA40942-GT AA40942-FL
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 20 mesh woven from 0.382mm (0.015in) Type 304
Stainless steel
Open area: 49%; Width of opening: 0.035in

Uses: Filtration, electrodes


Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty