Would you like to log in again?
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Diameter 0.51 mm (0.02 in) Type 304
Supplier: Thermo Scientific Chemicals
Danger
40946-BZ 40946-BW 040946-BW 040946-BZ
AA40946-BWEA 34.09 USD InStock
CAAA40946-BW AA40946-BW CAAA40946-BZ AA40946-BZ
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Diameter 0.51 mm (0.02 in) Type 304
Stainless steel
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty