Would you like to log in again?
Glassy Carbon Electron Beam Evaporating Crucibles, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40963-KT 040960-KT 040965-KT 040961-KT 40962-KT 40964-KT
AA40964-KTEA 780.93 USD InStock
AA40964-KT AA40961-KT AA40960-KT AA40965-KT AA40963-KT AA40962-KT
Glassy Carbon Electron Beam Evaporating Crucibles, Thermo Scientific
Crucibles
Glassy carbon offers high purity, corrosion resistance, thermal stability, and a structure impermeable to both gases and liquids
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
All vessels feature a 15° angle. These crucibles are designed for high-temperature differential thermal/thermal gravimetric analyses.
Alta Aesar's glassy carbon products are subjected to a high temperature heat treatment which imparts special material properties, including significantly improved corrosion resistance and strength.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty