Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.5%, Environmental Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid ethyl esterAcetic etherAcetidinAcetoxyethaneEthyl acetic esterEthyl ethanoateVinegar naphtha

040977-M1 40977-M1 040977-K2 040977-K7 40977-K7 40977-K2
AA40977-K2EA 56 USD InStock
CAAA40977-M1 AA40977-K2 AA40977-M1 CAAA40977-K2 CAAA40977-K7 AA40977-K7
Ethyl acetate ≥99.5%, Environmental Grade
Ethyl acetate
Pesticide residue, environmental, and GC analysis


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C
Melting Pt: –84 °C
Density: 0.902
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Color (APHA) 10
H2O 0.0005
Evaporation residue 5 ppm
Halogenated residue 5 ppt GC-ECD
Note: Submicron filtered

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty