Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.8%, Environmental Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol

040980-M6 040980-K2 40980-K2 40980-M6 40980-K7 040980-K7
AA40980-K2EA 45.59 USD InStock
CAAA40980-K2 AA40980-K2 CAAA40980-M6 AA40980-K7 AA40980-M6 CAAA40980-K7
Methanol ≥99.8%, Environmental Grade
Methanol
Suitable for environmental, pesticide, and GC analysis


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C
Melting Pt: –98 °CC
Density: 0.791
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty