Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Diameter 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

41039-G6 41039-G6 41039-G5 41039-G1 41039-G5 41039-G1
AA41039-G5EA 193.99 USD InStock
CAAA41039-G6 AA41039-G5 AA41039-G6 CAAA41039-G5 AA41039-G1 CAAA41039-G1
Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Diameter 0.05 mm (0.002 in)
Tungsten (gold coated)


Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty