Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium disulfate tetrahydrate 98+% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

41041-36 41041-22 041041-22 041041-A3 41041-A3 041041-36
AA41041-36EA 142.91 USD InStock
AA41041-36 CAAA41041-22 AA41041-22 CAAA41041-A3 CAAA41041-36 AA41041-A3
Zirconium disulfate tetrahydrate 98+% (metals basis)
Zirconium disulphate tetrahydrate
Catalyst support
Soluble in water


Formula: O₈S₂Zr·4H₂O
MW: 355.396 g/mol
Density: 3,22
Storage Temperature: Ambient
MDL Number: MFCD00149267
CAS Number: 7446-31-3
EINECS: 238-694-4
Merck Index: 14,10182
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty