Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodiphenylphosphine 98+%, rods

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DPPC

041352-18 41352-30 41352-18 041352-09 41352-09 041352-30
AA41352-30EA 342.99 USD InStock
CAAA41352-18 CAAA41352-30 AA41352-30 AA41352-18 CAAA41352-09 AA41352-09
Chlorodiphenylphosphine 98+%, rods
Chlorodiphenylphosphine


Formula: C₁₂H₁₀ClP
MW: 220.64 g/mol
Boiling Pt: 318…321 °C
Melting Pt: 14…16 °C
Density: 1,204
Flash Pt: 138 °C (280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000529
CAS Number: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty