Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl acetate 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid allyl ester

041353-AD 41353-AD 41353-AK 041353-AK
AA41353-ADEA 30.49 USD InStock
AA41353-AD CAAA41353-AD CAAA41353-AK AA41353-AK
Allyl acetate 99%
Allyl acetate


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 103…104 °C
Melting Pt: 6 °C
Density: 0,928
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008721
CAS Number: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3,II
Merck Index: 12,00293
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty