Synonyms:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II)
041359-04
41359-02
41359-09
41359-04
041359-02
041359-09
AA41359-02EA
70.19
USD
InStock
CAAA41359-02
CAAA41359-04
AA41359-02
AA41359-04
CAAA41359-09
AA41359-09
Bis(2,2,6,6-tetramethyl-2,5-heptanedionato-O,O')cobalt(II) ≥99.9% (metals basis)
Bis(2,2,6,6-tetramethyl-2,5-heptanedionato-O,O')cobalt(II)
Precursor for CVD process
Decomposes at 250°C