Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroiodomethane 97+%

Supplier: Thermo Scientific Chemicals
Warning

041479-14 41479-14 041479-22 41479-22
AA41479-14EA 387.99 USD InStock
CAAA41479-22 CAAA41479-14 AA41479-14 AA41479-22
Trifluoroiodomethane 97+%
Trifluoroiodomethane


Formula: CF₃I
MW: 195.91 g/mol
Boiling Pt: –22.5 °C
Melting Pt: –110 °C
Density: 2,361
Storage Temperature: Ambient
MDL Number: MFCD00001060
CAS Number: 2314-97-8
EINECS: 219-014-5
UN: 1956
ADR: 2.2,
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty