Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Di-n-octyl phthalate 98%

Supplier: Thermo Scientific Chemicals

41522-AE 41522-AP 41522-AC 041522-AP 041522-AE 041522-AC
AA41522-AEEA 163.99 USD InStock
AA41522-AE CAAA41522-AC AA41522-AP AA41522-AC CAAA41522-AE CAAA41522-AP
Di-n-octyl phthalate 98%
Di-n-octyl phthalate
Applications: Plasticizer


Formula: C₂₄H₃₈O₄
Boiling Pt: 380 °C
Melting Pt: –25 °C
Density: 0,98
Flash Pt: 219 °C (426 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015292
CAS Number: 117-84-0
EINECS: 204-214-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty