Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium hydride ≥92%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Calcium dihydride

41590-36 41590-22 041590-22 041590-36
AA41590-22EA 173.81 USD InStock
AA41590-22 CAAA41590-36 CAAA41590-22 AA41590-36
Calcium hydride ≥92%
Calcium hydride


Formula: H₂Ca
MW: 42.09 g/mol
Melting Pt: 600 °C (dec.)
Density: 1,900
Storage Temperature: Ambient
MDL Number: MFCD00010897
CAS Number: 7789-78-8
EINECS: 232-189-2
UN: 1404
ADR: 4.3,I
Merck Index: 13,01674
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty