Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ditungsten pentaboride ≥99.4% (metals basis)

Supplier: Thermo Scientific Chemicals
Synonyms: Tungsten boride

041664-14 041664-22 41664-14 41664-22
AA41664-14EA 65.49 USD InStock
AA41664-14 CAAA41664-14 AA41664-22 CAAA41664-22
Ditungsten pentaboride ≥99.4% (metals basis)
Ditungsten pentaboride


Formula: B₅W₂
Storage Temperature: Ambient
MDL Number: MFCD00049707
CAS Number: 12007-98-6
EINECS: 234-524-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty