Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triisopropylphosphine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tri-i-propylphosphine

41684-09 041684-04 041684-09 41684-04
AA41684-09EA 824.99 USD InStock
AA41684-09 CAAA41684-09 AA41684-04 CAAA41684-04
Triisopropylphosphine 98%
Triisopropylphosphine


Formula: C₉H₂₁P
MW: 160.24 g/mol
Boiling Pt: 176…178 °C
Density: 0.839
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015029
CAS Number: 6476-36-4
EINECS: 229-333-1
UN: 2845
ADR: 4.2,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty