Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Styrene-4-sulfonic acid sodium salt

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium-p-styrenesulphonateSodium 4-vinylbenzenesulphonate4-Vinylbenzenesulfonic acid sodium saltp-Styrenesulfonic acid sodium saltp-Styrenesulphonic acid sodium saltSodium-p-styrenesulfonate4-Vinylbenzenesulphonic acid sodium saltSodium 4-vinylbenzenesulfonate

41690-18 41690-30 041690-30 041690-18
AA41690-18EA 85.68 USD InStock
CAAA41690-18 AA41690-18 AA41690-30 CAAA41690-30
Styrene-4-sulfonic acid sodium salt
Styrene-4-sulfonic acid sodium salt


Formula: C₈H₇NaO₃S
MW: 206.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013379
CAS Number: 2695-37-6
EINECS: 220-266-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty