Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Multi Ion Cation IC Standard Solution, Specpure® (Dionex), Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific
Specpure®
041695-AE 41695-AE
AA41695-AEEA 275.21 USD InStock
AA41695-AE CAAA41695-AE
Multi Ion Cation IC Standard Solution, Specpure® (Dionex), Thermo Fisher Scientific
Standards Cationic Standards Multi-Element Standards
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD03792456

Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD03792456

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty